2-Amino-5-bromo-6-fluorobenzonitrile

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

023916-25G 023916-5G
FLUO023916-25GEA 0 GBP
FLUO023916-25G FLUO023916-5G
2-Amino-5-bromo-6-fluorobenzonitrile
2-Amino-5-bromo-6-fluorobenzonitrile
Formula: C₇H₄BrFN₂
MDL Number: MFCD06657979
CAS Number: 845866-92-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR