4-Bromo-N-butyl-3-methoxybenzamide

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

CA-4634-5G CA-4634-1G
COBBCA-4634-5GEA 0 GBP
COBBCA-4634-5G COBBCA-4634-1G
4-Bromo-N-butyl-3-methoxybenzamide
4-Bromo-N-butyl-3-methoxybenzamide
Formula: C₁₂H₁₆BrNO₂
MDL Number: MFCD11053852
CAS Number: 1065074-09-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR