3-Bromo-N-tert-butyl-4-methoxybenzamide

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

CA-4802-1G CA-4802-25G CA-4802-5G
COBBCA-4802-1GEA 0 GBP
COBBCA-4802-1G COBBCA-4802-25G COBBCA-4802-5G
3-Bromo-N-tert-butyl-4-methoxybenzamide
3-Bromo-N-tert-butyl-4-methoxybenzamide
Formula: C₁₂H₁₆BrNO₂
MDL Number: MFCD02655677
CAS Number: 356550-24-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR