3-Bromo-N-ethyl-5-nitrobenzamide

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

CA-4355-5G CA-4355-1G
COBBCA-4355-5GEA 0 GBP
COBBCA-4355-5G COBBCA-4355-1G
3-Bromo-N-ethyl-5-nitrobenzamide
3-Bromo-N-ethyl-5-nitrobenzamide
Formula: C₉H₉BrN₂O₃
MDL Number: MFCD09258762
CAS Number: 929000-30-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR