3-Bromo-N-isopropyl-5-nitrobenzamide

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

CA-4374-5G CA-4374-1G
COBBCA-4374-5GEA 0 GBP
COBBCA-4374-5G COBBCA-4374-1G
3-Bromo-N-isopropyl-5-nitrobenzamide
3-Bromo-N-isopropyl-5-nitrobenzamide
Formula: C₁₀H₁₁BrN₂O₃
MDL Number: MFCD09475855
CAS Number: 941294-16-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR