4-Bromo-N-ethylbenzamide

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

CA-4425-5G CA-4425-25G CA-4425-1G
COBBCA-4425-5GEA 0 GBP
COBBCA-4425-5G COBBCA-4425-25G COBBCA-4425-1G
4-Bromo-N-ethylbenzamide
4-Bromo-N-ethylbenzamide
Formula: C₉H₁₀BrNO
MDL Number: MFCD01211865
CAS Number: 41882-25-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR