4-Bromo-N-ethyl-3-methylbenzamide
Supplier: COMBI-BLOCKS
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
CA-4567-5G
CA-4567-1G
COBBCA-4567-5GEA
0
GBP
COBBCA-4567-5G
COBBCA-4567-1G
4-Bromo-N-ethyl-3-methylbenzamide
4-Bromo-N-ethyl-3-methylbenzamide
Formula:
C₁₀H₁₂BrNO |
MDL Number:
MFCD09972131 CAS Number: 1020252-82-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...