3-Bromo-N-ethylbenzamide
Supplier: COMBI-BLOCKS
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
CA-4479-1G
CA-4479-5G
COBBCA-4479-1GEA
0
GBP
COBBCA-4479-1G
COBBCA-4479-5G
3-Bromo-N-ethylbenzamide
3-Bromo-N-ethylbenzamide
Formula:
C₉H₁₀BrNO MW: 228.0900 g/mol |
MDL Number:
MFCD01215180 CAS Number: 26819-10-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...