2-Bromo-N-pentylbenzamide

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

CS-9420-5G CS-9420-1G
COBBCS-9420-5GEA 0 GBP
COBBCS-9420-5G COBBCS-9420-1G
2-Bromo-N-pentylbenzamide
2-Bromo-N-pentylbenzamide
Formula: C₁₂H₁₆BrNO
MDL Number: MFCD01212204
CAS Number: 349092-69-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR