2-Bromo-N-ethylbenzamide

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

CS-9407-5G CS-9407-1G
COBBCS-9407-5GEA 0 GBP
COBBCS-9407-5G COBBCS-9407-1G
2-Bromo-N-ethylbenzamide
2-Bromo-N-ethylbenzamide
Formula: C₉H₁₀BrNO
MDL Number: MFCD01211894
CAS Number: 80031-02-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR