4-Bromo-N-(prop-2-ynyl)benzamide

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 4-Bromo-N-(prop-2-yn-1-yl)benzamide

CA-4417-5G CA-4417-1G
COBBCA-4417-5GEA 0 GBP
COBBCA-4417-5G COBBCA-4417-1G
4-Bromo-N-(prop-2-ynyl)benzamide
4-Bromo-N-(prop-2-ynyl)benzamide
Formula: C₁₀H₈BrNO
MW: 238.0800 g/mol
MDL Number: MFCD09800901
CAS Number: 82225-32-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR