3-Bromo-N,N-dimethyl-5-nitrobenzamide
Supplier: COMBI-BLOCKS
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
CA-4353-5G
CA-4353-1G
COBBCA-4353-5GEA
0
GBP
COBBCA-4353-5G
COBBCA-4353-1G
3-Bromo-N,N-dimethyl-5-nitrobenzamide
3-Bromo-N,N-dimethyl-5-nitrobenzamide
Formula:
C₉H₉BrN₂O₃ |
MDL Number:
MFCD09258761 CAS Number: 929000-26-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...