3-Bromo-N,N-diethyl-5-nitrobenzamide

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

CA-4358-5G CA-4358-1G
COBBCA-4358-5GEA 0 GBP
COBBCA-4358-5G COBBCA-4358-1G
3-Bromo-N,N-diethyl-5-nitrobenzamide
3-Bromo-N,N-diethyl-5-nitrobenzamide
Formula: C₁₁H₁₃BrN₂O₃
MDL Number: MFCD09258763
CAS Number: 929000-34-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR