4-Bromo-N,N-diethylbenzamide

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

CA-4427-100G CA-4427-5G CA-4427-1G CA-4427-25G
COBBCA-4427-100GEA 0 GBP
COBBCA-4427-100G COBBCA-4427-5G COBBCA-4427-1G COBBCA-4427-25G
4-Bromo-N,N-diethylbenzamide
4-Bromo-N,N-diethylbenzamide
Formula: C₁₁H₁₄BrNO
MW: 256.1390 g/mol
MDL Number: MFCD00463697
CAS Number: 5892-99-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR