4-Bromo-N,N-diisopropylbenzamide
Supplier: COMBI-BLOCKS
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
CA-4413-1G
CA-4413-25G
CA-4413-5G
COBBCA-4413-1GEA
0
GBP
COBBCA-4413-1G
COBBCA-4413-25G
COBBCA-4413-5G
4-Bromo-N,N-diisopropylbenzamide
4-Bromo-N,N-diisopropylbenzamide
Formula:
C₁₃H₁₈BrNO MW: 284.1921 g/mol |
MDL Number:
MFCD00591594 CAS Number: 79606-46-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...