2-Bromo-N,N-diethylbenzamide

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

CS-9401-5G CS-9401-1G
COBBCS-9401-5GEA 0 GBP
COBBCS-9401-5G COBBCS-9401-1G
2-Bromo-N,N-diethylbenzamide
2-Bromo-N,N-diethylbenzamide
Formula: C₁₁H₁₄BrNO
MW: 256.1390 g/mol
MDL Number: MFCD00457139
CAS Number: 76041-86-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR