3-Bromo-N-(3-chloropropyl)benzenesulphonamide

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

OT-0154-5G OT-0154-1G
COBBOT-0154-5GEA 0 GBP
COBBOT-0154-5G COBBOT-0154-1G
3-Bromo-N-(3-chloropropyl)benzenesulphonamide
3-Bromo-N-(3-chloropropyl)benzenesulphonamide
Formula: C₉H₁₁BrClNO₂S
MW: 312.6111 g/mol
MDL Number: MFCD09258645
CAS Number: 929000-46-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR