2-Bromo-N-(4-chlorophenyl)benzamide

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

CS-9432-5G CS-9432-1G
COBBCS-9432-5GEA 0 GBP
COBBCS-9432-5G COBBCS-9432-1G
2-Bromo-N-(4-chlorophenyl)benzamide
2-Bromo-N-(4-chlorophenyl)benzamide
Formula: C₁₃H₉BrClNO
MDL Number: MFCD00442557
CAS Number: 66569-06-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR