5-Bromo-2-chloro-N-methylbenzamide

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

CA-4717-1G CA-4717-5G CA-4717-25G
COBBCA-4717-1GEA 0 GBP
COBBCA-4717-1G COBBCA-4717-5G COBBCA-4717-25G
5-Bromo-2-chloro-N-methylbenzamide
5-Bromo-2-chloro-N-methylbenzamide
Formula: C₈H₇BrClNO
MDL Number: MFCD05853706
CAS Number: 435273-54-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR