5-Bromo-2-chloro-N-ethylbenzamide
Supplier: COMBI-BLOCKS
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
CA-4716-5G
CA-4716-25G
CA-4716-1G
COBBCA-4716-5GEA
0
GBP
COBBCA-4716-5G
COBBCA-4716-25G
COBBCA-4716-1G
5-Bromo-2-chloro-N-ethylbenzamide
5-Bromo-2-chloro-N-ethylbenzamide
Formula:
C₉H₉BrClNO |
MDL Number:
MFCD04219747 CAS Number: 150079-92-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...