2-Methoxy-5-(methoxycarbonyl)phenylboronic acid

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 2-Methoxy-5-Methoxycarbonylphenyboronic acid, (2-Methoxy-5-(methoxycarbonyl)phenyl)boronic acid

BB-5015-1G BB-5015-5G
COBBBB-5015-1GEA 0 GBP
COBBBB-5015-1G COBBBB-5015-5G
2-Methoxy-5-(methoxycarbonyl)phenylboronic acid
2-Methoxy-5-(methoxycarbonyl)phenylboronic acid
Formula: C₉H₁₁BO₅
MDL Number: MFCD09800877
CAS Number: 221006-63-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR