4-(Cyclohexyl(methyl)carbamoyl)phenylboronic acid

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: (4-(Cyclohexyl(methyl)carbamoyl)phenyl)boronic acid

BB-3506-5G BB-3506-1G
COBBBB-3506-5GEA 0 GBP
COBBBB-3506-5G COBBBB-3506-1G
4-(Cyclohexyl(methyl)carbamoyl)phenylboronic acid
4-(Cyclohexyl(methyl)carbamoyl)phenylboronic acid
Formula: C₁₄H₂₀BNO₃
MW: 261.1300 g/mol
MDL Number: MFCD09027224
CAS Number: 913835-84-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR