3-(Diethylcarbamoyl)phenylboronic acid ≥98%
Supplier: Thermo Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Warning
|
Synonyms:
3-(N,N-Diethylaminocarbonyl)phenylboronic acid, 3-(Diethylcarbamoyl)benzeneboronic acid, (3-(Diethylcarbamoyl)phenyl)boronic acid
H53061.03
H53061.06
H53061.03EA
0
GBP
H53061.03
H53061.06
3-(Diethylcarbamoyl)phenylboronic acid ≥98%
3-(Diethylcarbamoyl)phenylboronic acid
Purity: 98%
CAS No.: 237413-05-7
Molecular Formula: C11H16BNO3
Formula Weight: 221.06
Melting Point: 132-136°
MDL No.: MFCD04115684
Formula:
C₁₁H₁₆BNO₃ MW: 221.06 g/mol Melting Pt: 132…136 °C Storage Temperature: Ambient |
MDL Number:
MFCD04115684 CAS Number: 237413-05-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...