(4-Bromo-2-nitrophenyl)acetic acid

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 4-Bromo-2-nitrophenylacetic acid

AS21766G1 AS21766G5
ACTIAS21766G1EA 61 GBP
ACTIAS21766G1 ACTIAS21766G5
(4-Bromo-2-nitrophenyl)acetic acid
(4-Bromo-2-nitrophenyl)acetic acid
Formula: C₈H₆BrNO₄
MW: 260.0400 g/mol
MDL Number: MFCD05663950
CAS Number: 6127-11-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR