(2-Bromo-6-nitrophenyl)acetic acid

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

AS23206G1 AS23206G5
ACTIAS23206G1EA 207 GBP
ACTIAS23206G1 ACTIAS23206G5
(2-Bromo-6-nitrophenyl)acetic acid
(2-Bromo-6-nitrophenyl)acetic acid
Formula: C₈H₆BrNO₄
MDL Number: MFCD10566470
CAS Number: 37777-74-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR