(2-Bromo-5-chlorophenyl)acetic acid

Supplier: ACTIVATE SCIENTIFIC GMBH
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 2-(2-Bromo-5-chlorophenyl)acetic acid

ACTIAS25079G25EA 107 GBP
ACTIAS25079G25 ACTIAS25079G100
(2-Bromo-5-chlorophenyl)acetic acid
(2-Bromo-5-chlorophenyl)acetic acid
Formula: C₈H₆BrClO₂
MW: 249.4900 g/mol
MDL Number: MFCD00153890
CAS Number: 81682-38-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR