3-(Hexylamino)propionitrile

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

018707-10G 018707-25G
FLUO018707-10GEA 0 GBP
FLUO018707-10G FLUO018707-25G
3-(Hexylamino)propionitrile
3-(Hexylamino)propionitrile
Formula: C₉H₁₈N₂
MDL Number: MFCD00060315
CAS Number: 55490-85-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR