2-(2-Fluorophenyl)oxazole

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

HC-3303-5G HC-3303-1G
COBBHC-3303-5GEA 0 GBP
COBBHC-3303-5G COBBHC-3303-1G
2-(2-Fluorophenyl)oxazole
2-(2-Fluorophenyl)oxazole
Formula: C₉H₆FNO
MDL Number: MFCD15143568
CAS Number: 178672-06-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR