5-(4-Nitrophenyl)-4-oxazolecarboxylic acid

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

HC-6234-5G HC-6234-1G
COBBHC-6234-5GEA 0 GBP
COBBHC-6234-5G COBBHC-6234-1G
5-(4-Nitrophenyl)-4-oxazolecarboxylic acid
5-(4-Nitrophenyl)-4-oxazolecarboxylic acid
Formula: C₁₀H₆N₂O₅
MDL Number: MFCD08558969
CAS Number: 914220-30-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR