5-(4-Chlorophenyl)-4-oxazolecarboxylic acid

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

HI-1354-5G HI-1354-1G
COBBHI-1354-5GEA 0 GBP
COBBHI-1354-5G COBBHI-1354-1G
5-(4-Chlorophenyl)-4-oxazolecarboxylic acid
5-(4-Chlorophenyl)-4-oxazolecarboxylic acid
Formula: C₁₀H₆ClNO₃
MDL Number: MFCD00105450
CAS Number: 143659-14-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR