5-(3-Chlorophenyl)-4-oxazolecarboxylic acid
Supplier: COMBI-BLOCKS
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
HC-6233-5G
HC-6233-1G
COBBHC-6233-5GEA
0
GBP
COBBHC-6233-5G
COBBHC-6233-1G
5-(3-Chlorophenyl)-4-oxazolecarboxylic acid
5-(3-Chlorophenyl)-4-oxazolecarboxylic acid
Formula:
C₁₀H₆ClNO₃ |
MDL Number:
MFCD01934492 CAS Number: 255876-54-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...