Methyl-5-(4-fluorophenyl)-4-oxazolecarboxylate
Supplier: COMBI-BLOCKS
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
HC-6254-5G
HC-6254-1G
COBBHC-6254-5GEA
0
GBP
COBBHC-6254-5G
COBBHC-6254-1G
Methyl-5-(4-fluorophenyl)-4-oxazolecarboxylate
Methyl 5-(4-fluorophenyl)-4-oxazolecarboxylate
Formula:
C₁₁H₈ClNO₃ |
MDL Number:
MFCD09800994 CAS Number: 89204-90-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...