Methyl-5-(2-chlorophenyl)-4-oxazolecarboxylate

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

HC-6256-5G HC-6256-1G
COBBHC-6256-5GEA 0 GBP
COBBHC-6256-5G COBBHC-6256-1G
Methyl-5-(2-chlorophenyl)-4-oxazolecarboxylate
Methyl 5-(2-chlorophenyl)-4-oxazolecarboxylate
Formula: C₁₁H₈ClNO₃
MDL Number: MFCD09878374
CAS Number: 89204-91-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR