5-(4-Bromophenyl)oxazole

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

AS20799G1 AS20799G25 AS20799G5
ACTIAS20799G1EA 26 GBP
ACTIAS20799G1 ACTIAS20799G25 ACTIAS20799G5
5-(4-Bromophenyl)oxazole
5-(4-Bromophenyl)oxazole
Formula: C₉H₆BrNO
MW: 224.0540 g/mol
MDL Number: MFCD02682015
CAS Number: 72571-06-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR