4-Methyl-2-(2-nitrophenyl)oxazole

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

HI-1331-1G HI-1331-250M
COBBHI-1331-1GEA 0 GBP
COBBHI-1331-1G COBBHI-1331-250M
4-Methyl-2-(2-nitrophenyl)oxazole
4-Methyl-2-(2-nitrophenyl)oxazole
Formula: C₁₀H₈N₂O₃
MDL Number: MFCD09801027
CAS Number: 951884-48-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR