5-(2,4-Dichlorophenyl)-4-(hydroxymethyl)oxazole

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

HC-6368-1G HC-6368-250M
COBBHC-6368-1GEA 0 GBP
COBBHC-6368-1G COBBHC-6368-250M
5-(2,4-Dichlorophenyl)-4-(hydroxymethyl)oxazole
5-(2,4-Dichlorophenyl)-4-(hydroxymethyl)oxazole
Formula: C₁₀H₇Cl₂NO₂
MDL Number: MFCD09878381
CAS Number: 957062-73-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR