2-Amino-4-thiazolecarboxylic acid

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 2-Aminothiazole-4-carboxylic acid

HI-1166-5G HI-1166-25G
COBBHI-1166-5GEA 0 GBP
COBBHI-1166-5G COBBHI-1166-25G
2-Amino-4-thiazolecarboxylic acid
2-Amino-4-thiazolecarboxylic acid
Formula: C₄H₄N₂O₂S
MW: 144.15 g/mol
MDL Number: MFCD00859429
CAS Number: 40283-41-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR