Ethyl thiazole-4-carboxylate

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

HI-1120-5G HI-1120-1G
COBBHI-1120-5GEA 0 GBP
COBBHI-1120-5G COBBHI-1120-1G
Ethyl thiazole-4-carboxylate
Ethyl thiazole-4-carboxylate
Formula: C₆H₇NO₂S
MW: 157.1900 g/mol
MDL Number: MFCD03788566
CAS Number: 14527-43-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR