Ethyl-2-amino-5-oxazolecarboxylate

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: Ethyl 2-aminooxazole-5-carboxylate

HI-1198-1G HI-1198-25G HI-1198-5G
COBBHI-1198-1GEA 0 GBP
COBBHI-1198-1G COBBHI-1198-25G COBBHI-1198-5G
Ethyl-2-amino-5-oxazolecarboxylate
Ethyl 2-amino-5-oxazolecarboxylate
Formula: C₆H₈N₂O₃
MW: 156.1393 g/mol
MDL Number: MFCD06657433
CAS Number: 113853-16-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR