Ethyl-5-oxazolecarboxylate

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: Ethyl oxazole-5-carboxylate

HI-1250-5G HI-1250-1G
COBBHI-1250-5GEA 0 GBP
COBBHI-1250-5G COBBHI-1250-1G
Ethyl-5-oxazolecarboxylate
Ethyl 5-oxazolecarboxylate
Formula: C₆H₇NO₃
MW: 141.13 g/mol
MDL Number: MFCD04114930
CAS Number: 118994-89-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR