5-Amino-3-(4-fluorophenyl)isoxazole

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

AS23795G1 AS23795G5
ACTIAS23795G1EA 209 GBP
ACTIAS23795G1 ACTIAS23795G5
5-Amino-3-(4-fluorophenyl)isoxazole
5-Amino-3-(4-fluorophenyl)isoxazole
Formula: C₉H₇FN₂O
MDL Number: MFCD03407398
CAS Number: 81465-82-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR