3-(4-Chlorophenyl)isoxazole
Supplier: COMBI-BLOCKS
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
HC-6993-1G
HC-6993-5G
COBBHC-6993-1GEA
0
GBP
COBBHC-6993-1G
COBBHC-6993-5G
3-(4-Chlorophenyl)isoxazole
3-(4-Chlorophenyl)isoxazole
Formula:
C₉H₆ClNO |
MDL Number:
MFCD08446383 CAS Number: 31301-39-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...