5-Amino-3-(2-bromophenyl)isoxazole

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

AS23804G1 AS23804G5
ACTIAS23804G1EA 0 GBP
ACTIAS23804G1 ACTIAS23804G5
5-Amino-3-(2-bromophenyl)isoxazole
5-Amino-3-(2-bromophenyl)isoxazole
Formula: C₉H₇BrN₂O
MDL Number: MFCD07383024
CAS Number: 119162-51-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR