5-(4-Bromophenyl)isoxazole

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

HC-6884-5G HC-6884-25G HC-6884-1G
COBBHC-6884-5GEA 0 GBP
COBBHC-6884-5G COBBHC-6884-25G COBBHC-6884-1G
5-(4-Bromophenyl)isoxazole
5-(4-Bromophenyl)isoxazole
Formula: C₉H₆BrNO
MDL Number: MFCD02183532
CAS Number: 7064-31-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR