5-(3-Bromophenyl)isoxazole
Supplier: COMBI-BLOCKS
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
HC-6885-5G
HC-6885-1G
COBBHC-6885-5GEA
0
GBP
COBBHC-6885-5G
COBBHC-6885-1G
5-(3-Bromophenyl)isoxazole
5-(3-Bromophenyl)isoxazole
Formula:
C₉H₆BrNO MW: 224.06 g/mol |
MDL Number:
MFCD02183531 CAS Number: 7064-33-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...