Ethyl-5-methylisoxazole-3-carboxylate 95%

Supplier: Thermo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: Ethyl 5-methylisoxazole-3-carboxylate

A13226.06 A13226.03
A13226.06EA 94 GBP
A13226.06 A13226.03
Ethyl-5-methylisoxazole-3-carboxylate 95%
Ethyl 5-methylisoxazole-3-carboxylate

Purity: 95% CAS No.: 3209-72-1 Molecular Formula: C7H9NO3 Formula Weight: 155.15 Flash Point: 109°(228°F) Melting Point: 27-31° Boiling Point: 77-79°/0.5mm MDL No.: MFCD00085080

Formula: C₇H₉NO₃
MW: 155.15 g/mol
Boiling Pt: 77…79 °C (0.5 mmHg)
Melting Pt: 27…31 °C
Flash Pt: 109 °C (228 °F)
Storage Temperature: Ambient
MDL Number: MFCD00085080
CAS Number: 3209-72-1
EINECS: 221-720-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR