4-Bromoisoxazole

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

HI-1452-5G HI-1452-1G
COBBHI-1452-5GEA 0 GBP
COBBHI-1452-5G COBBHI-1452-1G
4-Bromoisoxazole
4-Bromoisoxazole
Formula: C₃H₂BrNO
MW: 147.9600 g/mol
MDL Number: MFCD02179569
CAS Number: 97925-43-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR