3-Iodo-5-methoxy-1H-indazole
Supplier: COMBI-BLOCKS
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
HI-1605-1G
HI-1605-250M
COBBHI-1605-1GEA
0
GBP
COBBHI-1605-1G
COBBHI-1605-250M
3-Iodo-5-methoxy-1H-indazole
3-Iodo-5-methoxy-1H-indazole
Formula:
C₈H₇IN₂O |
MDL Number:
MFCD07781661 CAS Number: 290367-99-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...