5-Amino-3-iodo-1H-indazole

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 3-Iodo-1H-indazol-5-amine

AS12489G1 AS12489G5
ACTIAS12489G1EA 102 GBP
ACTIAS12489G1 ACTIAS12489G5
5-Amino-3-iodo-1H-indazole
5-Amino-3-iodo-1H-indazole
Formula: C₇H₆IN₃
MW: 259.0471 g/mol
MDL Number: MFCD07781656
CAS Number: 599183-36-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR