3-Amino-6-fluoro-1H-indazole

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

AS22561G1 AS22561G5
ACTIAS22561G1EA 0 GBP
ACTIAS22561G1 ACTIAS22561G5
3-Amino-6-fluoro-1H-indazole
3-Amino-6-fluoro-1H-indazole
Formula: C₇H₆FN₃
MDL Number: MFCD06739129
CAS Number: 404827-75-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR